Learn More
Coverguard™ FFP1 NR D Face Mask
Product Code. 12023186
Change view
Click to view available options
Style:
Flat Foldable Half-Mask
Flat Foldable Half-Mask with Valve
Shell Mask
Shell Mask with Valve
Nosepiece Type:
Adjustable Nose Clip
Adjustable Nose Clip with PU Foam Nose Pad
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 120
Pack of 240
Product Code. Style Nosepiece Type With Exhalation Valve unitSize
12023186 Flat Foldable Half-Mask with Valve Adjustable Nose Clip Yes Pack of 240
12003186 Flat Foldable Half-Mask Adjustable Nose Clip No Pack of 240
12013186 Shell Mask Adjustable Nose Clip with PU Foam Nose Pad No Pack of 240
12033186 Shell Mask with Valve Adjustable Nose Clip with PU Foam Nose Pad Yes Pack of 120
4 options
Product Code. 12023186
Brand: Coverguard™ 23105
Only null left
Add to basket
Add to basket
The Coverguard™ FFP1 masks are single-use and protect against non-toxic solid and liquid aerosols, in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
  • PPE Cat. III FFP1 NR D
  • Disposable
  • Electrostatic filter media
  • Sliding elastic band for better adjustment
  • Latex-free, non-woven polypropylene
  • Adjustable nose clip
Specifications
Certifications/Compliance (Europe) CE PPE Category III FFP1 NR D, EN 149:2001 + A1:2009
Description FFP1 Disposable Foldable Half-Mask, Sealed Outer Contour, Exhalation Valve
Disposable Single-use
Material Non-woven Polypropylene
With Exhalation Valve Yes
Filter Efficiency FFP1
Latex Free Yes
Material Headstrap Synthetic Rubber Elastic (Latex Free)
Nosepiece Type Adjustable Nose Clip
Resistant To Non-Toxic Aerosols and Non-Toxic Dusts
Sterility Non-sterile
Style Flat Foldable Half-Mask with Valve