Learn More
Coverguard™ FFP2 NR D Face Mask
Description
- PPE Cat. III FFP2 NR D
- Disposable
- Electrostatic filter media
- Sliding elastic band for better adjustment
- Latex-free, non-woven polypropylene
- Adjustable nose clip
Specifications
Specifications
| Certifications/Compliance (Europe) | CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009 |
| Description | FFP2 Disposable Half Mask, Sealed Outer Contour, Exhalation Valve |
| Disposable | Single-use |
| With Exhalation Valve | Yes |
| For Use With (Equipment) | Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure |
| Nosepiece Type | Adjustable Nose Clip |
| Resistant To | Non-Toxic Solid and Liquid Aerosols |
| Sterility | Non-sterile |
| Style | Half Mask with Exhalation Valve |