Learn More
Coverguard™ FFP2 NR D Face Mask for Organic Vapors Gas and Acids
Click to view available options
:
Face Mask; Style: Half Mask with Shell and Carbon Valve
Description
- PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
- Filters bad odors
- Exhalation valve with active carbon reduces fog build-up inside the mask
- Non-woven polypropylene
- Adjustable aluminum nose clip with PU foam total nose seal
- Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Specifications
Specifications
| Certifications/Compliance (Europe) | CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009 |
| Description | FFP2 NR D Half Mask with Valve for Organic Vapors, Gas and Acids |
| Disposable | Single-use |
| With Exhalation Valve | Yes |
| For Use With (Equipment) | Heavy & Process Industries, Chemical and Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY |
| Nosepiece Type | Aluminum Nose Clip, Total PU Nose Seal |
| Resistant To | Organic Vapors, Gas and Acids |
| Shape | Half Mask |
| Sterility | Non-sterile |
| Style | Half Mask with Shell and Carbon Valve |