Learn More
Coverguard™ FFP2 NR D SL VO Face Mask for Organic Vapors Gas and Acids with Exhalation Valve
FFP2 half masks have an electrostatic filter media to filter bad odors. The design gives limited breathing resistance, thanks to the filtering material and exhalation valve with active carbon, aids breathing and avoids fog build-up inside the mask.
Brand: Coverguard™ 23238
Additional Details : Weight : 0.01000kg
Beschreibung
- PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
- Filters bad odors
- Exhalation valve with active carbon reduces fog build-up inside the mask
- Non-woven polypropylene
- Adjustable aluminum nose clip with PU foam total nose seal
- Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Spezifikation
CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009 | |
Single-use | |
Heavy & Process Industries, Chemical & Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY | |
Aerosols and Low Toxic Dust, Organic Vapors, Gas and Acids | |
Non-sterile |
FFP2 Half Mask with Shell and Carbon Valve | |
Yes | |
Aluminum Adjustable Nose Clip, PU Foam Nose Pad | |
Half Mask |