missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human 17 beta-HSD1/HSD17B1 ELISA Kit (Colorimetric)

Product Code. 18674020 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18674020

Brand: Novus Biologicals™ NBP260473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.781 to 50 ng/mL (example only, lot dependent)
Assay Sensitivity 0.5 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3292
Gene Symbol HSD17B1
Quantity 1 Kit
Research Discipline Breast Cancer, Cancer
Storage Requirements Storage is content dependent.
Synonym 17-beta-HSD 1, 17-beta-hydroxysteroid dehydrogenase type 1, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E17KSR, E2DH, EC 1.1.1.62, EDH17B1, EDH17B2EDHB17, estradiol 17-beta-dehydrogenase 1, estradiol 17-beta-dehydrogenase-1, HSD17, hydroxysteroid (17-beta) dehydrogenase 1, hydroxysteroid (17-beta) dehydrogenase 1 isoform, MGC138140, Placental 17-beta-hydroxysteroid dehydrogenase, SDR28C1, short chain dehydrogenase/reductase family 28 CE, member 1
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.