Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
Brand:  Thermo Scientific™ 010-1072
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15332167
  • 915.00€ / Pack of 2
Estimated Shipment: 24-06-2024
to see stock.
Description
Description
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
Specifications
Specifications
30mL Oak Ridge Tube
2