Learn More
BRAND™ Sterile Filling Tubes for seripettor™ Bottle Top Dispensers
Product Code. 10101681 Shop All Brand Products
Click to view available options
For Use With (Equipment):
Seripettor™ Bottletop Dispenser
Unit Size:
Each
Product Code. 10101681
Brand: BRAND™ 704538
Only null left
Add to basket
Add to basket
Includes additional O-ring. Autoclavable for sterile applications.
  • For use with seripettor™ bottletop dispensers
  • Polypropylene construction
  • Autoclavable for sterile applications
  • Includes an additional O-ring
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With (Equipment) Seripettor™ Bottletop Dispenser
Includes O-ring
Quantity 1 Ea.
Sterility Sterile
Length (Metric) 500 mm