Learn More
BRAND™ Sterile Filling Tubes for seripettor™ Bottle Top Dispensers
Shop All Brand Products
Click to view available options
For Use With (Equipment):
Seripettor™ Bottletop Dispenser
Unit Size:
Each
Description
- For use with seripettor™ bottletop dispensers
- Polypropylene construction
- Autoclavable for sterile applications
- Includes an additional O-ring
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Equipment) | Seripettor™ Bottletop Dispenser |
| Includes | O-ring |
| Quantity | 1 Ea. |
| Sterility | Sterile |
| Length (Metric) | 500 mm |