Learn More
Sperian™ Honeywell™ Filter Canisters for Respirators
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Filter Canisters for Respirators have low positioning so as not to obstruct the field of vision.
Brand:  Sperian™ 1001619
Additional Details : Weight : 1.61100kg
Product Code. 11827161
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • User checks the proper installation via the audible click on its safety system
  • Coconut based carbon for filtration
  • Mechanical filtration membrane
Specifications
Specifications
Canister
Brown
Filter Canister
Half-Masks and Full-Face Masks
0.8 kg
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 1141, EC Attestation- EC Attestation, EC Attestation Number- 1141, Certifications- EN 1438
Filters against gas and organic vapours
Respiratory Protection
10/Pk.