missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Filter Disc
Brand: Haldenwanger™ 31B/22
Additional Details : Weight : 0.00200kg
Specifications
For Use with 35mL Porcelain Gooch Crucibles | |
10Pack | |
Filter Disc | |
22 mm |