Learn More
LNI Swissgas Filter Elements for NG CASTORE XS iQ
Filter elements for NG CASTORE XS iQ.
Brand: LNI Swissgas 6930.00.170
This item is not returnable.
View return policy
Description
- Replace every 4000 hours or 1 year.
Specifications
Filter Element |