Learn More
LNI Swissgas Filter Elements for NG CASTORE XS iQ
Filter elements for NG CASTORE XS iQ.
Brand:  LNI Swissgas 6930.00.170
Product Code. 17872189
  • 352.00€ / Each
Description
  • Replace every 4000 hours or 1 year.
Specifications
Specifications
Filter Element