Learn More
ROBU VitraPOR™ Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
Brand: ROBU VitraPOR™ 21110
Additional Details : Weight : 0.10000kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
1000 mL | |
Buchner Shape | |
1 Ea. |
Borosilicate Glass 3.3 | |
0 |