missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Filter Funnels

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07195470

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10040431

  • 285.00€ / Each

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

  • Filter funnels
  • Material: borosilicate glass 3.3
Specifications

Specifications

1 L
320 mm
120 mm
Borosilicate Glass 3.3
120 mm
4
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions