Learn More
Witeg™ Filter Funnels
Witeg™ Filter Funnels include a filter disc for separating materials from liquid solutions.
Brand:  Witeg™ 2 367 124
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11807820
  • 53.80€ / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass construction
  • Variety of volumetric capacities available
  • Variety of disc porosities available
  • Variety of stem sizes available
Specifications
Specifications
Filter funnel
125 mL
60mm
60mm
10mm
4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers