Learn More
Cytiva Filter Holder
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Brand:  Cytiva 10461200
Additional Details : Weight : 0.59000kg
 View more versions of this product
Product Code. 11944556
  • 394.00€ / Pack of 5
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Specifications
Specifications
FP050/0 PSU
Polysulfone
5
50 mm