Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
Brand:  Thermo Scientific™ 1425505
Additional Details : Weight : 0.02700kg
 View more versions of this product
Product Code. 11709508
  • 262.00€ / Each
In Stock
to see stock.
Description
Description
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Specifications
Optical Filter
Multiskan FC
Multiskan™
550 nm