missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Filtrode Electrode

Brand:  Hamilton™ 242064

Additional Details : Weight : 0.13800kg

Product Code. 10469894

  • 257.00€ / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0 ± 20mV
12 mm
Use this electrode for your toughest applications.
0 to 14
S7 Electrical Connector
Filtrode Electrode
120 mm
0°C to 60°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions