Learn More
Schneider Gerd™ Fine Blunt Tip Laboratory Forceps
Click to view available options
Length (Metric):
130 mm
145 mm
Tip Style:
Fine blunt, bent
Fine, Blunt
Unit Size:
Each
Description
- Fine Blunt Tip Laboratory Forceps
- Ideal for dissection and general lab applications
Specifications
Specifications
| Tip Style | Fine blunt, bent |
| Length (Metric) | 130 mm |
| Product Type | Forceps |
| Quantity (No. per Case) | Each |
| Type | Forceps, Laboratory |