Learn More
Schneider Gerd™ Fine Blunt Tip Laboratory Forceps
Use these forceps for a variety of applications. Schneider Gerd™ Fine Blunt Tip Laboratory Forceps feature a grooved body for optimal grip.
6.35€ - 18.90€
Specifications
Product Type | Forceps |
---|---|
Quantity (No. per Case) | Each |
Type | Forceps, Laboratory |
Product Code | Brand | Tip Style | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | Length (Metric) | Price | Quantity & Availability | ||||
10326504
|
Schneider Gerd™
8132 |
Fine, Blunt | 130 mm |
6.35€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
10480914
|
Schneider Gerd™
8142 |
Fine blunt, bent | 130 mm |
18.90€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
10440914
|
Schneider Gerd™
8113 |
Fine, Blunt | 145 mm |
6.45€
Each |
In Stock
Log in to see stock. |
||||
Description
- Fine Blunt Tip Laboratory Forceps
- Ideal for dissection and general lab applications
Specifications
Forceps | |
Each | |
Forceps, Laboratory |