missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Finedex™ Latex Gloves
Click to view available options
Size:
9
10
Description
- Natural latex glove for flexibility, dexterity, and good mechanical and chemical resistance against detergents.
- Cotton flock interior for comfort.
- Diamond-pattern finish for grasping dry and damp objects.
- Cuff protects forearm.
- Minimum latex protein.
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374-2003 Chemical hazards, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0462, EC Attestation Number 0075/797/162/11/08/0462 |
Classification | Dexterity: 5 |
Color | Yellow |
Cut Resistance Rating | 0 |
Exterior Finish | Diamond |
Material | Latex, Cotton |
Product Type | Gloves |
Size | 9 |
Thickness (Metric) | 0.4 to 0.8 mm |
For Use With (Application) | Building, Construction, Catering, Chemical, Fishing, Food, Industrial Cleaning, Laboratory, Medical, Pharmaceutical, Printing, Ship Building and Water Treatment industries |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'