Learn More
Esculape Laboratoire™ First Aid Kit Refill
Product Code. p-7217837
Click to view available options
Sufficient For:
10 Persons
8 People
Unit Size:
Each
Product Code. 11718403
Brand: Esculape Laboratoire™ 805 05 05
Only null left
Add to basket
Add to basket
Refill kit for Esculape Laboratoire first aid kit
  • Refill kit
  • For Esculape Laboratoire first aid kit
Specifications
For Use With (Application) First Aid Kit
Product Type Refill Kit
Sufficient For 8 People