missing translation for 'onlineSavingsMsg'
Learn More

Erlab BF2 FILTER - BE

Product Code. 18666458
Click to view available options
For Use With (Equipment):
Fume Hood
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18666458

Brand: Erlab H01012202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filter for your filtering storage cabinet 632A/832BF.

  • Filter for your filtering storage cabinet 632A/832BF
  • Type of filter: Molecular (acid vapours)
  • Dimensions: 380 x 404 x 62 mm
  • Quantity/Set: 1

Specifications

For Use With (Equipment) Fume Hood
Product Type Carbon Filter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.