Learn More
Fisher BioReagents™ Microbiology Media Additives: Casamino Acids
Additive for preparing bacterial nutrient media
Brand:  Fisher BioReagents™ 12747109
Additional Details : CAS : 9000-71-9 Weight : 0.53910kg
 View more versions of this product
Product Code. 12747109
  • 501.00€ / 500g
Estimated Shipment: 23-06-2021
Add to basket
Description
Description
Dehydrated.
Specifications
Specifications
Molecular genetics
Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production
Powder
Casamino Acid
500g
Poly Bottle