Learn More
Fisher BioReagents™ SOB Broth (Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Brand: Fisher BioReagents™ BP9737-500
Additional Details : CAS : 8013-01-2 Weight : 0.50000kg
Description
- Directions for Media Preparation: Add 2 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat and frequently agitate to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 14g of broth
Specifications
Capsule | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures | |
36 x 14 g |
SOB Broth | |
Casein Peptone 20g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L, Magnesium Sulfate 2.4g/L, Potassium Chloride 0.186g/L | |
Poly Bottle |