Learn More
Fisherbrand™ Technical Pens
Product Code. 15203976 Shop All Fisherbrand Products
Click to view available options
Color:
Green
Red
Width (Metric) Line:
0.2 mm
0.4 mm
Unit Size:
Pack of 6
Product Code. 15203976
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams.
Fisher Scientific™ Technical Pens are ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams. 6 per pack.
Available in multiple colors and widths.
Specifications
Tip Style Fine
Product Type Technical Pen
Color Red
For Use With (Application) Ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams
Quantity 6 Pk.
Width (Metric) Line 0.4 mm