Learn More
Fisherbrand™ 10-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
Shop All Thermo Scientific Products
Click to view available options
Thickness (English):
0.8mm
1.5mm
Unit Size:
Each
Description
- For use with Owl P2-CST Multiple Gradient Gel Caster
Specifications
Specifications
| For Use With (Equipment) | Owl P2-CST Multiple Gradient Gel Caster |
| Type | Comb |
| Gel Dimensions | 20 x 20 cm |
| Teeth Count | 10 |
| Thickness (English) | 0.8mm |
| Thickness (Metric) Teeth | 0.08 cm |
| Volume (Metric) Well | 239 μL |
| Length (English) Footprint | 9.84 in. |
| Length (English) Gel | 7.8 in. |
| Length (Metric) Footprint | 25 cm |
| Show More |