Learn More
Fisherbrand™ 3 Tier Disc Assembly Complete
Shop All Fisherbrand Products
Click to view available options
Includes:
12 Clips
24 Clips
Unit Size:
Each
Description
- Rotary Mixer Disc complete with 12 clips for tubes, supplied unassembled
- Suitable for 'End to End Mixing' tubes
- Clips for Tube diameters between 15 - 17 mm that are suitable for 15 mL Centrifuge tubes
- The range of accessories can also be autoclaved and allow for easy loading and tube removal.
- The accessory is easy to fit onto the RM-1 or RM-2 locating on three pins then secured in place with thumb wheel
Specifications
Specifications
| Includes | 24 Clips |