Learn More
Fisherbrand™ 3 Tier Disc Assembly Complete
Product Code. p-8015454 Shop All Fisherbrand Products
Click to view available options
Includes:
12 Clips
24 Clips
Unit Size:
Each
Product Code. 12388448
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for 'End to End Mixing' tubes, these clips for tube diameters between 15 - 17mm, are ideal for 15ml Centrifuge tubes.
  • Rotary Mixer Disc complete with 12 clips for tubes, supplied unassembled
  • Suitable for 'End to End Mixing' tubes
  • Clips for Tube diameters between 15 - 17mm that are suitable for 15ml Centrifuge tubes
  • The range of accessories can also be autoclaved and allow for easy loading and tube removal.
  • The accessory is easy to fit onto the RM-1 or RM-2 locating on three pins then secured in place with thumb wheel
Specifications
Includes 12 Clips