Learn More
Fisherbrand™ 96-Well Polystyrene Plates
Shop All Fisherbrand Products
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 180
Pack of 80
Description
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment
- Polystyrene
- Plates stack easily without lids
- Available in flat or round well shapes
- Nonsterile
- Untreated, provides uniform surface properties
- Use 12-565-511 lid (sold separately)
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Round Bottom (U-well) |
| For Use With (Application) | Ideal for antibiotic screens, cell based assays and screening compounds |
| Quantity | 180/Cs. |