Learn More
Fisherbrand™ Cryogenic Vial Storage Boxes
GREENER_CHOICE
Product Code. 12902431 Shop All Fisherbrand Products
Click to view available options
Holds:
1.2 to 2 mL Threaded Cryo Vials
5 mL Cryo Vials
Unit Size:
Pack of 24
Pack of 4
Product Code. 12902431
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
  • 81-Place white polycarbonate storage boxes
  • Use in mechanical freezers and liquid nitrogen Dewars
  • Cover is printed with a 9 x 9 numbered grid for inventory purposes
  • Stackable
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Color White
Material Polycarbonate
Shape Square
Type Cryo Storage Box
For Use With (Application) For ultra-low temperature storage of both internally and externally threaded cyrogenic vials
Holds 1.2 to 2 mL Threaded Cryo Vials
Tube Array 9 x 9
Dimensions (L x W x H) 5.25 x 5.25 x 2 in. (13.3 x 13.3 x 5 cm)
Length (English) 5.25 in.
Length (Metric) 13.3 cm
Width (English) 5.25 in.
Width (Metric) 13.3 cm
Height (English) 2 in.
Height (Metric) 5 cm
Weight (Metric) 4.31 kg