Learn More
Fisherbrand™ Natural HDPE Round Handled Jug with White Polypropylene PE Foam Cap
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Pack of 40
Description
- Feature finger grips for easy handling
- Provides moisture barrier and are impact resistant
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Capacity (English) | 64 oz. |
| Color | Natural |
| Material | HDPE |
| Quantity | 40/Cs. |
| Closure Color | White |
| Liner Material | PE Foam |
| Neck Finish | 38-400 |
| Mouth | Narrow |
| Shape | Round |
| Show More |