Learn More
Fisherbrand™ Suprasil™ Quartz Fluorescence Precision Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Fisherbrand™ Suprasil™ Quartz Flourescence Precision Cell Cuvettes are supplied with a PTFE stopper.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Sold in pairs
- Very chemically resistant
- Maintains shape even at high temperatures up to approximately 1000°C
- 80% transmission from 320 to 2500nm