Learn More
Hellma Suprasil™ Quartz Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Suprasil™ Quartz Micro Cell Cuvettes are supplied with a PTFE cover.
331.00€ - 515.00€
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11784716
|
Hellma
104002B-10-40 |
Suprasil quartz windows and black quartz sidewalls |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
11771784
|
Hellma
104-002-10-40 |
Suprasil quartz windows; Height: 45mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Sold in pairs
- Very chemically resistant
- Maintains shape, even at high temperatures up to approximately 1000°C
- 80% transmission from 320 to 2500nm
Specifications
Quartz |