Learn More
Fisherbrand™ Rotary Mixer Discs
Product Code. 12387488 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
15-17 mm dia. Tubes
8-11 mm dia. Tubes
Unit Size:
Each
Product Code. 12387488
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for end-to-end mixing tubes
  • Rotary Mixer Disc complete with clips for tubes, supplied unassembled
  • Suitable for 'End to End Mixing' tubes,
  • Clips for tube diameters between  8 - 11 mm are suitable for 0.5mL, 1.5mL and 2.0mL Microcentrifuge tubes, 3mL Vacutainers and 5mL capped tubes (8 -11 mm)
  • Clips for tube diameters between 15 - 17 mm are suitable for 15 mL centrifuge tubes
  • Easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
Product Type Rotary Mixer Disc
Description Rotary Mixer Disc
For Use With (Equipment) 15-17 mm dia. Tubes