Learn More
Fisherbrand™ Silicone Recessed Septum Stoppers
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
13.75 to 14.25 mm
14.50 to 15.50 mm
20.00 to 21.00 mm
9.22 to 9.75 mm
Unit Size:
Pack of 10
Specifications
Specifications
| Diameter (Metric) Inner | 20.00 to 21.00 mm |
| For Use With (Application) | Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal |
| For Use With (Equipment) | For 20 to 21mm I.D. containers |
| Size | 3 |
| Color | Translucent |
| Material | Silicone |
| Product Type | Septum Stopper |
| Stopper Number | 3 |