Learn More
Fisherbrand™ SureGrip™ Pipet Tip Racked or Reload System Tips
Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks
128.66€
Specifications
Sterility Non-sterile
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
View More Specs
Products 1
Product Code Brand Volume (Metric) Tip Style Format Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Tip Style Format Color Price Quantity & Availability  
11837393
View Documents Promotion Details
Fisherbrand™
1250 μL Standard Length Refill Natural This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
Full line of disposable pipet tips available in preloaded carriers for fast, easy loading into empty racks
Easy-to-use: just load the carrier into an empty pipet tip rack (reuse your old racks or purchase separately). Carrier locks onto rack with a quick, press-and-snap action. When empty, recycle or dispose of the carrier and reuse the rack.
  • Compatible with most manufacturers' racks
  • Reduces waste by 80% Certified DNase, RNase, Human DNA and PCR inhibitor free
Pipet Tips in Racks
  • All tips are available racked in sturdy, reusable CellRacks
  • Withstand numerous autoclave cycles
  • Keep tips aligned for pipetter mounting
Several Tip Sizes Available
  • 10μL Pipet tip has graduation marks at 2μL and 10μL
  • 10μL Long Reach tip features extended length (45mm) to reach the bottom of tubes without contaminating pipetter shaft, and graduation marks at 2μL, 5μL and 10μL
  • 200μL Pipet tip features a soft-fit collar for easy mounting and ejection, and graduation marks at 10μL, 50μL and 100μL
  • 1250μL Pipet tip features an extended length (103mm), racked on 9mm centers (multichannel compatible), and graduation marks at 200μL, 500μL and 1000μL
Order Info
Each pack consists of ten preloaded tip carriers of 96 tips each.
Specifications
Specifications
Non-sterile
Single-use
Yes
DNase, RNase, DNA, and PCR inhibitor Free
Universal Fit
SureGrip