Learn More
Microspec™ Fixed Volume Pipette
Single channel pipette with fixed volume
Brand:  Microspec™ AD6
Additional Details : Weight : 0.06800kg
 View more versions of this product
Product Code. 16645462
  • 110.00€ / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Fixed volume
  • Single channel
  • Non-sterile
  • Grey
Specifications
Specifications
0.01
Gray
Non-sterile
100 μL
Not Autoclavable
1
Fixed Volume Pipette