Learn More
BOLA Flanging Tips
Product Code. 15432404
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1.6 mm
2.4 mm
Unit Size:
Each
Product Code. 15432404
Brand: BOLA F70954
Only null left
Add to basket
Add to basket
Flanging Tips for making flanges. Suitable for BOLA Thermoelectric Flanging Tools F 701-12 and F 701-14.
For flanging different inner diameters of tubing, suitable for BOLA Thermo-electric Flanging Tools
Specifications
Diameter (Metric) Inner 1.6 mm
Autoclavable Autoclavable