Learn More
Grant Instruments™ Multi-level Flat Platform Tower With Non-Slip Mat for PSU20i
Click to view available options
Description:
2-level
3-level
4-level
Unit Size:
Each
Description
- For use with PSU 20i Orbital Shaking Platform
- 2, 3, or 4 levels
- Black rubber pads provide nonslip surface for vessels on the platform.
Specifications
Specifications
| Material | Rubber |
| Dimensions (L x W) | 480 x 380 mm |
| Description | 4-level |
| For Use With (Equipment) | PSU-20i |
| No. of Shelves | 4-level |