Learn More
Proteintech FlexAble HRP Antibody Labeling Kit for Human IgG
Shop All Proteintech Products
Click to view available options
Quantity:
10 Tests
100 Tests
50 Tests
Unit Size:
10 reactions
200 reactions
50 reactions
Description
FlexAble HRP Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate HRP in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifications
Specifications
| Quantity | 10 Tests |
| Format | Liquid |