Learn More
Sartorius Flexible Sample Holder, for Cubis™ Balances
Product Code. 11777624 Shop All Sartorius Products
Click to view available options
:
Cubis, Q-grip
Product Code. 11777624
Brand: Sartorius YFH01MS
For use with Cubis Balances. Q-Grip is a flexible and adaptable one-size-fits-all holder for bottles, test tubes, reaction containers or filters (up to 120mm) for all Cubis semi-microbalances and analytical balances.
Includes:
  • Plate
  • Filter supports (2)
  • Flask supports (2)
  • Pan support
  • Clamping screw and nut
Replaces the original weighing pan and is only for CUBIS Balances with 0.1 and 0.01mg readability.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Containers and Filters of Cubis Semi-microbalance
Type Weighting Flask Holder