Learn More
Key Instruments™ Flo Rite™ FR2000 Model Clear Acrylic Flowmeter with Standard Inlet Valve
Click to view available options
Airflow Rate:
1 to 10 LPM
10 to 100 LPM
2 to 25 LPM
4 to 50 LPM
Unit Size:
Each
Specifications
Specifications
| Type | Flowmeter |
| Accuracy | ±5% |
| Airflow Rate | 4 to 50 LPM |
| Float Material | Stainless Steel |
| Pressure Rating | 100 psig |
| Connections | 1/8 in. NPT (F) |
| For Use With (Application) | Air sampling equipment aquaculture, Desalinization equipment, Gas analyzers, Medical systems, Photoprocessing equipment, Water treatment and distribution systems |
| Height (Metric) | 101.6 mm |
| Internal Part Material | Acrylic |
| Model | FR2000 |
| Show More |