Learn More
Semadeni™ Floating Ball-Type Waste Disposal Unit
The need to operate a lid is eliminated, without allowing vapors or odors to escape from the waste container.
Brand:  Semadeni™ 4770
Additional Details : Weight : 0.50000kg
Product Code. 12826370
  • 151.00€ / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • With screw-on funnel with self-closing, patented floating ball valve
  • With additional overflow protection and splashguard
  • Removable filter on the filling tube to retain solid matter (glass shards, magnetic stirrers, etc.)
  • Canisters stand in sturdy polypropylene containment trays, capable of holding the entire canister contents in the event of an emergency
  • 1 can, 1 x 6 L bottles
  • 250 x 250 x 440 mm
Specifications
Specifications
Waste Disposal Unit
1/Pk.