Learn More
Thermo Scientific™ Fluophase™ PFP Javelin™ Guard Columns
Product Code. 11553321 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
20 mm
Unit Size:
Pack of 4
Product Code. 11553321
Brand: Thermo Scientific™ 82705022106
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Particle size: 5μm
Features of Fluophase PFP (Perfluorophenyl) Solid Phase
  • Perfluorinated phases provide unique selectivity
  • Extra retention and selectivity for halogenenated compounds
  • Excellent for taxane analysis
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase PFP