Learn More
Fluopyram, Fluka™
Product Code. 16272912 Shop All Honeywell Fluka Products
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize'
50mg
Product Code. 16272912
missing translation for 'mfr': Honeywell-Fluka R163350MG
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Analytical standard, reference material
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fluopyram
Packaging Glass Bottle
CAS 658066-35-4
Melting Point 119°C
Boiling Point Decomposes below the boiling point.
Synonym N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide
Industry Type Environmental Analysis
Molecular Weight (g/mol) 396.71
Quantity 50 mg
Grade Analytical Standard