missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Free Acid, Reag. Ph. Eur., Honeywell Fluka™

Product Code. p-7120436 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15623820

Brand: Honeywell-Fluka 32615100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reag. Ph. Eur., free acid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 314°C to 316°C
UN Number NONH for all modes of transport
Beilstein 94324
Formula Weight 332.31g/mol
Odor Odourless
Packaging Plastic Bottle
Suitability Passes test for UV spectrum
Color Index No. Acid Yellow 73
Color Brown
Quantity 100 g
Chemical Name or Material Fluorescein, Reag. Ph. Eur.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.