missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-Ubiquitin

Product Code. 11525016
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
This item is not returnable. View return policy

Product Code. 11525016

Brand: Invitrogen PV4377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescein-Ubiquitin

Fluorescein-labeled ubiquitin for use as a general research tool or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with FRET donors including LanthaScreen™ Tb-Ubiquitin (PV4375) or a LanthaScreen™ antibody including Tb-anti-GST (PV4216) or Tb-anti-ubiquitin-FK2 (PV4751).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg
Conjugate Fluorescein
Content And Storage Fluorescein-labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340⁄490, 520
For Use With (Equipment) Microplate Reader
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes
Readout End Point
Shipping Condition Dry Ice
Substrate Properties Protein-Based Substrate
Technique TR-FRET
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.