Learn More
Novus Biologicals™ Fluorescent Particle Array (Carboxyl, Odd # peaks) Kit
Product Code. 18629874 Shop All Bio Techne Products
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
1 set
Product Code. 18629874
Brand: Novus Biologicals™ NBP318591
Used to develop assays for allergy testing, autoimmune diseases, cardiac markers, cytokine detection, endocrine markers, infectious disease markers, isotyping, genotyping, kinase and phosphorylated protein activity, metabolic markers, and tissue typing
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. Do not freeze.
For Use With (Application) Flow Cytometry
Format Deionized water
Preservative 0.02% Sodium Azide
Quantity 5 x 1 mL