Learn More
Memmert™ Flush-Fit Sets without Air Slots for UN30
Product Code. 17510066 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Product Code. 17510066
Brand: Memmert™ UN30B29729
Only null left
Add to basket
Add to basket
Stainless steel frame covering gap between oven and wall opening.
  • Stainless steel
  • Without air slots
  • Techincal clarification required
Specifications
For Use With (Equipment) UN30 Oven
Type Flush-fit Unit
Material Stainless Steel