Learn More
Eurofins DiscoverX Fms Protein, Active
Descripción
CSF1R (colony stimulating factor I receptor), formerly known as FMS (McDonough feline sarcoma viral oncogene), is a receptor tyrosine kinase for CSF-1 and expressed primarily in monocytes, macrophages, and human tumor tissue. CSF-1 binding to FMS induces dimerization of the tyrosine kinase domain of the receptor, and stimulates the survival, proliferation, and differentiation of haematopoietic cells of the monocyte-macrophage series.
GenBank U63963
Especificaciones
Especificaciones
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 50.2 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥75% |
| Protein | Fms, active |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.