Learn More
BTX™ Gemini X2 Electroporation Generator Footswitch
Allows for hands free operation of Gemini X2, ECM2001+, ECM830 and ECM630 generator (touch screen models). Harvard Apparatus Gemini X2 Electroporation Footswitch functions as the start button on front of the generator.
Brand: BTX™ 452030
Additional Details : Weight : 0.51000kg
Description
- Allows for hands free operation of Gemini X2 generator
- Desirable when conducting in vivo/in ovo gene delivery or nuclear transfer/cloning when both hands are needed for sample manipulation
- Functions as start button on front of the generator
Specifications
Hands free operation of electroporation generators | |
BTX Electroporation | |
3000 VDC, 250 VAC | |
1 Ea. | |
Electrode Connector Cable | |
AC Sine Wave, DC Square Wave |
Gemini X2, ECM 2001+, ECM 830, and ECM 630 Foot Pedal (compatible with touchscreen models) | |
Electroporation accessory | |
Gemini X2, ECM 2001+, ECM 830, and ECM 630 Foot Pedal | |
Non-sterile | |
Footswitch |